missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic acid dihydrate, Honeywell Fluka™
missing translation for 'orderingAttributeHoverText'
Quantità:
500 g
1 kg
5 kg
25 kg
Beschreibung
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation
Percent Purity | 99.5% (min.) |
Grado | ACS |
Quantità | 5 kg |
Perdita all'essiccatura | 28 - 30 % |
Imballaggio | Plastic Bottle |
Forma fisica | Powder |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'