missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2RY13 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20335-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP.Specifica
P2RY13 | |
Polyclonal | |
Unconjugated | |
P2RY13 | |
FKSG77, GPCR1, GPR86, GPR94, P2RY13, P2Y13, SP174 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53829, 74191 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BPV8, Q9D8I2 | |
P2RY13 | |
P2RY13 Fusion Protein Ag14108 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |