Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Union Tee
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Marca: Cole-Parmer™ 31320-29
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Superior chemical resistance with durable PFA composition.
- Get a tight seal at higher pressures.
- Withstand higher pressures than barbed fittings.
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifica
Compression Union Tee | |
Autoclavable | |
1/8 in. |
Compression Union Tee | |
PFA | |
Autoclave or Ethylene Oxide |