missing translation for 'onlineSavingsMsg'
Learn More

PCDH11Y Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 20069-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899544

  • 140.00€ / 20µL
Spedizione stimata: 15-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The antibody can recognize PCDH11X and PCDH11Y.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its most closely related cadherin superfamily member is located in this homologous region on the X chromosome. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with N- and C-terminal variation.
Specifica

Specifica

PCDH11Y
Polyclonal
Unconjugated
PCDH11Y
PCDH PC, PCDH Y, PCDH11, PCDH11X, PCDH11Y, PCDH22, PCDHY, Protocadherin 11, protocadherin 11 Y linked, Protocadherin 22, Protocadherin PC, Protocadherin prostate cancer
Rabbit
Antigen Affinity Chromatography
RUO
27328, 83259
-20°C
Liquid
Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BZA7, Q9BZA8
PCDH11X, PCDH11Y
Peptide
20 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali