missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDHX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10951-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The PDHX gene encodes component X of the pyruvate dehydrogenase (PDH) complex. For a detailed description of the pyruvate dehydrogenase complex. The mammalian PDH complex differs from that in E. coli and from the other mammalian alpha-keto acid dehydrogenases by the presence of a 53-kDa protein called protein X. Component X binds to the E3 component of the PDH complex (Robinson et al., 1990 ; Aral et al., 1997 ).Specifica
PDHX | |
Polyclonal | |
Unconjugated | |
PDHX | |
DLDBP, E3 binding protein, E3BP, OPDX, PDHX, PDX1, proX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27402, 311254, 8050 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00330, Q8BKZ9 | |
Pdhx | |
PDHX Fusion Protein Ag1389 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |