Learn More
Masterflex™ PFA Compression Threaded Adapter, Male NPT
Superior Compression Threaded Adapter are chemical resistance with durable PFA
Marca:  Masterflex™ 31321-63
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 12354538
  • 67.20€ / Pezzo
Spedizione stimata: 26-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifica
Specifica
Vacuum Tubing
4 mm
Autoclave or ethylene oxide (ETO)
Compression Size / Tube Od (Port 1): 4 mm
PFA compression threaded adapter, 4 mm tubing OD x 1/4 in. male NPT
Compression x NPT(M)
1/4 in.
PFA