missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11033-2-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the prefoldin beta subunit family. The encoded protein is one of six subunits of prefoldin, a molecular chaperone complex that binds and stabilizes newly synthesized polypeptides, thereby allowing them to fold correctly. The complex, consisting of two alpha and four beta subunits, forms a double beta barrel assembly with six protruding coiled-coils.Specifica
PFDN1 | |
Polyclonal | |
Unconjugated | |
PFDN1 | |
PDF, PFD1, PFDN1, prefoldin subunit 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5201 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60925 | |
PFDN1 | |
PFDN1 Fusion Protein Ag1506 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |