missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-BLNK (Tyr84) Recombinant Rabbit Monoclonal Antibody (BLNKY84-H4), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537085
Dettagli aggiuntivi : Peso : 0.01000kg
Description
B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.Specifications
Phospho-BLNK (Tyr84) | |
Monoclonal | |
PE | |
BLNK | |
AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa | |
Rabbit | |
Protein A/G | |
RUO | |
29760 | |
4° C | |
Liquid |
Flow Cytometry | |
BLNKY84-H4 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
Q8WV28 | |
BLNK | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr84 of human phospho BLNK. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG κ |