missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-BLNK (Tyr84) Recombinant Rabbit Monoclonal Antibody (BLNKY84-H4), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
185.00€ - 465.00€
Specifica
Antigene | Phospho-BLNK (Tyr84) |
---|---|
Clone | BLNKY84-H4 |
Applicazioni | Flow Cytometry |
Classificazione | Monoclonal |
Coniugato | PE |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17116928
|
Thermo Scientific
MA537335 |
10 Tests |
185.00€
pezzi 10 |
Spedizione stimata: 29-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17116678
|
Thermo Scientific
MA537085 |
100 Tests |
465.00€
pezzi 100 |
Spedizione stimata: 29-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.Specifica
Phospho-BLNK (Tyr84) | |
Flow Cytometry | |
PE | |
Rabbit | |
Human | |
Q8WV28 | |
29760 | |
IgG κ | |
Protein A/G | |
Antibody |
BLNKY84-H4 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.20% BSA and 0.09% sodium azide | |
AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa | |
BLNK | |
Primary | |
4° C | |
BLNK |