missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Chk1 (Ser317) Recombinant Rabbit Monoclonal Antibody (Chk1S317-F10), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537004
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-Chk1 (Ser317) Recombinant Monoclonal antibody specifically detects Phospho-Chk1 (Ser317) in Human, Mouse samples. It is validated for Flow Cytometry.Specifica
Phospho-Chk1 (Ser317) | |
Monoclonal | |
APC | |
Chek1 | |
C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W | |
Rabbit | |
Protein A/G | |
RUO | |
1111, 12649 | |
4° C | |
Liquid |
Flow Cytometry | |
Chk1S317-F10 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
O14757, O35280 | |
Chek1 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser317 of human phospho Chk1. | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |