Learn More
Phospho-STK11 (Thr189) Rabbit, anti-Human, Unconjugated, Clone: 5J9, Proteintech
Rabbit Recombinant Monoclonal Antibody
Marca: Proteintech 80127-1-RR-20UL
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
STK11(serine/threonine-protein kinase 11) is also named as LKB1, PJS, and belongs to the protein kinase superfamily. It controls the activity of AMP-activated protein kinase (AMPK) family members, thereby playing a role in various processes such as cell metabolism, cell polarity, apoptosis and DNA damage response. The tumour suppressor protein LKB1 is a serine/threonine kinase that has been causally linked to Peutz-Jeghers syndrome (PJS). Defects in STK11 are a cause of Peutz-Jeghers syndrome (PJS) and defects in STK11 have been associated with testicular germ cell tumor (TGCT) and some sporadic cancers, especially lung cancers.
Specifica
Phospho-STK11 (Thr189) | |
Monoclonal | |
250 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |
Western Blot, ELISA | |
5J9 | |
Unconjugated | |
20 μL | |
6794 | |
Liquid |