missing translation for 'onlineSavingsMsg'
Learn More

Pikachurin Mouse anti-Human, Clone: 1C6D12, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66232-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897915

  • 325.38€ / 150µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This monoclonal antibody detects two bands of 111 kDa and 55-60 kDa. The lower-molecular-weight band has been reported and likely represents a fragment resulting from cleavage of Pikachurin.

Involved in both the retinal photoreceptor ribbon synapse formation and physiological functions of visual perception. Necessary for proper bipolar dendritic tip apposition to the photoreceptor ribbon synapse. Promotes matrix assembly and cell adhesiveness.
Specifica

Specifica

Pikachurin
Monoclonal
2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q63HQ2
EGFLAM
Pikachurin Fusion Protein Ag6362
150 μL
Primary
Human
Antibody
IgG1
Western Blot
1C6D12
Unconjugated
EGFLAM
Agrin like protein, AGRINL, AGRNL, EGFLAM, Pikachurin
Mouse
Protein G
RUO
133584
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali