Learn More
VistaLab™ Technologies Pipetter Nozzles
Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter
Marca: VistaLab™ Technologies 9057-1013
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Available in range of sizes
Specifica
Non-sterile | |
Adjustable Models 0.2 μL, Fixed Models 1 μL and 2 μL | |
5/Pk. | |
1 in. |
2 μL | |
Filter | |
2 in. | |
1 in. |