Learn More
VistaLab™ Technologies Pipetter Nozzles
Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter
Marca: VistaLab™ Technologies 9060-1010
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Available in range of sizes
Specifica
Non-sterile | |
0.5 to 20 μL Ovation™ BioNatural™ Multichannel Pipette | |
2 in. | |
5 in. |
0.5 to 20 μL | |
4/Box | |
2 in. |