Learn More
VistaLab™ Technologies Pipetter Nozzles
Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter
Marca: VistaLab™ Technologies 9057-1010
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Available in range of sizes
Specifica
Non-sterile | |
Electronic Single Channel Model 0.5 to 20 μL, Adjustable Model 1 to 10 μL and 2 to 20 μL, Fixed Models 3 to 20 μL | |
10/Box | |
3 in. |
2 to 20 μL | |
Internal aerosol/liquid barrier filter | |
5 in. | |
1 in. |