missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4E Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18088-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Calcium-dependent phospholipase A2 that selectively hydrolyzes glycerophospholipids in the sn-2 position.Specifica
PLA2G4E | |
Polyclonal | |
Unconjugated | |
PLA2G4E | |
phospholipase A2, group IVE, PLA2G4E, PLA2G4E Ens | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
123745, 296091, 329502 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3MJ16, Q50L42 | |
Pla2g4e | |
PLA2G4E Fusion Protein Ag12808 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |