missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAC8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12284-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
PLAC8 is a protein coding gene.Specifica
PLAC8 | |
Polyclonal | |
Unconjugated | |
PLAC8 | |
C15, onzin, PLAC8, placenta 8, Protein C15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231507, 360914, 51316 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JI48, Q9NZF1 | |
Plac8 | |
PLAC8 Fusion Protein Ag2939 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |