Learn More
Motic™ CCIS™ EC-H Plan Phase Objective, Negative Phase
CCIS™ EC-H Plan Phase Objective, Negative Phase is compatible with all BA series microscopes
Marca: Motic™ 1101001703411
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- For use with BA Series Microscopes
- EC-H Plan Phase
- Negative Phase
- Magnifications/Numerical Apertures available:
- 10X/0.25 (WD=17.4mm)
- 20X/0.45 (WD=0.9mm)
- 40X/0.65/S (WD=0.5mm)
- 100X/1.25/S-Oil (WD=0.15mm)
Specifica
CCIS, Plan Achromatic Phase Objective, Negative Phase | |
0.5 mm WD | |
40X | |
1 Ea. |
EC-H Plan Phase | |
BA Microscope Series | |
0.65 |