Learn More
Buerkle™ Plastic Desiccator
Desiccators made of plastic, break resistant, lightweight and suitable for vacuums. Stopcock suitable for vacuums up to 980 mbar. Base and desiccator plate made out of Polypropylene. Lid is crystal clear Polycarbonate with a non-return valve and a venting valve. O-ring seal made of Polychlorobutadiene-rubber. Continuous temperature max. +60 °C.
Marca: Buerkle™ 9791-0150
100.50 EUR valido fino al 2024-12-31
Usa il codice promo "23267" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.42550kg
Descrizione
- Hose connection fits hoses with inner Ø 8.5 mm
- Delivery incl. O-ring
- Perforated plate and additional O-rings available as accessories
Specifica
White | |
60°C | |
150 mm | |
175 mm |