missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10589-2-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the delta class of phospholipase C enzymes. Phospholipase C enzymes play a critical role in many cellular processes by hydrolyzing phosphatidylinositol 4,5-bisphosphate into two intracellular second messengers, inositol 1,4,5-trisphosphate and diacylglycerol. Expression of this gene may be a marker for cancer.Specifica
PLCD4 | |
Polyclonal | |
Unconjugated | |
PLCD4 | |
hPLCD4, Phospholipase C delta 4, phospholipase C, delta 4, PLC delta 4, PLCD4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140693, 18802, 84812 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q62711, Q8K3R3, Q9BRC7 | |
Plcd4 | |
PLCD4 Fusion Protein Ag0937 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |