missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLDN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10891-2-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene may play a role in intracellular vesicle trafficking. It interacts with Syntaxin 13 which mediates intracellular membrane fusion. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifica
PLDN | |
Polyclonal | |
Unconjugated | |
BLOC1S6 | |
BLOC1S6, PA, PALLID, Pallid protein homolog, Pallidin, pallidin homolog (mouse), PLDN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18457, 26258, 317630 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4V8A6, Q9R0C0, Q9UL45 | |
Bloc1s6 | |
PLDN Fusion Protein Ag1337 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |