Learn More
Memmert™ Prolunghe a innesto per forno
Use these tube extensions with Memmert ovens. Memmert™ Plug-in Tube Extensions for Ovens are designed for exhaust air ducting and connection by hose if necessary.
Marca: Memmert™ B29718
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm)
- Angled
- Only models U, I, S
Specifica
Models U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 1060 | |
Plug-In Tube Extension | |
57 (Inside) mm, 60.3 (Outside) mm |