Learn More
Ismatec™ Reducer Tubing Connectors, Plastic
Variously sized and shaped reducer tubing connectors. Ismatec™ Reducer Tubing Connectors, Plastic are available in different plastic compositions.
Marca: Ismatec™ ISM571A
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- Reducer tubing connector, with varying inlet and outlet diameters.
- Designed for Ismatec™ pump tubing.
- Available in different sizes, shapes, and compositions.
Specifica
Reducer Tube Connector | |
For 2.5 and 3.0mm I.D. Tubing | |
2.5mm and 3.0mm I.D. |
Polypropylene | |
10/Pk. | |
Straight |