Learn More
Porcelaines Avignon™ Mortaio in porcellana
Crush and grind laboratory substances into powders using the Avignon™ Porcelain Pestle, Along with the mortar, it is an integrated laboratory tool in the world of science and medicine;
Marca: Porcelaines Avignon™ 15009
Dettagli aggiuntivi : Peso : 1.54800kg
Descrizione
- Sturdy porcelain
- Used with mortar
- Effective for wide variety of laboratory crushing needs
Specifica
1000 mL | |
Mortaio | |
Mortar | |
1000mL |
180 mm | |
Porcelain | |
100 mm |