Learn More
Porcelaines Avignon™ Mortaio in porcellana
Crush and grind laboratory substances into powders using the Avignon™ Porcelain Pestle, Along with the mortar, it is an integrated laboratory tool in the world of science and medicine;
Marca: Porcelaines Avignon™ 15210
Dettagli aggiuntivi : Peso : 0.74000kg
Descrizione
- Sturdy porcelain
- Used with mortar
- Effective for wide variety of laboratory crushing needs
Specifica
1500 mL | |
Pestello | |
Mortar | |
1500mL |
210mm | |
Porcellana | |
110mm |