missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcelaines Avignon™ Mortaio in porcellana
Crush and grind laboratory substances into powders using the Avignon™ Porcelain Pestle, Along with the mortar, it is an integrated laboratory tool in the world of science and medicine;
Brand: Porcelaines Avignon™ 15003
Additional Details : Peso : 0.80000kg
Description
- Sturdy porcelain
- Used with mortar
- Effective for wide variety of laboratory crushing needs
Specifications
150 mL | |
Mortaio | |
Mortar | |
150 ml |
102 mm | |
Porcelain | |
60 mm |