missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcelaines Avignon™ Mortaio in porcellana
Crush and grind laboratory substances into powders using the Avignon™ Porcelain Pestle, Along with the mortar, it is an integrated laboratory tool in the world of science and medicine;
Marque: Porcelaines Avignon™ 15005
Informations supplémentaires : Peso : 3.96000kg
Description
- Sturdy porcelain
- Used with mortar
- Effective for wide variety of laboratory crushing needs
Spécification
250 mL | |
Mortaio | |
Mortar | |
250 ml |
128 mm | |
Porcelain | |
75mm |