missing translation for 'onlineSavingsMsg'
Learn More

PPARA Mouse anti-Human, Clone: 1G1E10, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66826-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899055

  • 399.00€ / 150µL
Spedizione stimata: 21-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family termed peroxisome proliferator activated receptors (PPAR′s). Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Studies indicate that PPAR′s are activated by peroxisome proliferators such as clofibric acid, nafenopin, and WY-14,643, as well as by some fatty acids. It has also been shown that PPAR′s can induce transcription of acyl coenzyme A oxidase & cytochrome P450 (CYP450) A6 through interaction with specific response elements. PPAR, like several other nuclear hormone receptors, heterodimerizes with retinoid X receptor (RXR) alpha.
Specifica

Specifica

PPARA
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q07869
PPARA
PPARA Fusion Protein Ag7896
150 μL
Primary
Human
Antibody
IgG1
Western Blot
1G1E10
Unconjugated
PPARA
hPPAR, NR1C1, PPAR, PPAR alpha, PPARA
Mouse
Protein G
RUO
5465
-20°C
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali