missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPP4C (Human) Recombinant Protein
Human PPP4C full-length ORF ( AAH01416, 1 a.a. - 307 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005531-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: MAEISDLDRQIEQLRRCELIKESEVKALCAKAREILVEESNVQRVDSPVTVCGDIHGQFYDLKELFRVGGDVPETNYLFMGDFVDRGFYSVETFLLLLALKVRYPDRITLIRGNHESRQITQVYGFYDECLRKYGSVTVWRYCTEIFDYLSLSAIIDGKIFCVHGGLSPSIQTLDQIRTIDRKQEVPHDGPMCDLLWSDPEDTTGWGVSPRGAGYLFGSDVVAQFNAANDIDMICRAHQLVMEGYKWHFNETVLTVWSAPNYCYRCGNVAAILELDEHLQKDFIIFEAAPQETRGIPSKKPVADYFL
Specifica
AAH01416 | |
protein phosphatase 4 (formerly X), catalytic subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPP4C | |
GST |
5531 | |
Wheat germ expression system | |
25 μg | |
PP4, PPH3, PPX | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |