Learn More
Pro-Lab Diagnostics™ Prolex™ Streptococcal Grouping Latex Kits
For grouping Lancefield betahaemolytic Streptococci
Marca: Pro-Lab Diagnostics™ PL036
Dettagli aggiuntivi : CAS : 26628-22-8 Peso : 0.80000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Descrizione
- Instant results obtained at room temperature
- Advanced extraction process using modified nitrous acid reagents
- Enhanced results with blue carboxylate-modified microparticles (CM-MP)
- As few as one colony required
- Colour coded reagents and vials for ease of selection
- Available individually
Specifica
2°C to 8°C | |
Group G Latex Reagent | |
3mL Blue Latex particles coated with purified Rabbit Antibodies | |
3 mL |