missing translation for 'onlineSavingsMsg'
Learn More

ATAGO™ Multi-Wavelength Abbe Refractometer, Model DR-M2/1550

The ATAGO™ Multi-Wavelength Abbe Refractometer, Model DR-M2/1550 measures refractive index or Abbe number (vd or ve) at different wavelengths ranging from 450 to 1550nm.

Marca:  ATAGO™ 1412


Dettagli aggiuntivi : Peso : 0.10000kg

Codice prodotto. 11456248

  • 17755.00€ / Pezzo
Spedizione stimata: 23-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello

Includes: Refractometer, Power supply unit, Monochromatic light source device, Near infrared ray viewer, Interference filter (589nm)

Descrizione

Descrizione

  • LCD digital display
  • Measument achieved by matching boundary line with intersection point of cross hairs
  • Can be calibrated with water
  • Can be connected to printer and circulating

Measurement Range
Refractive index : 1.3278 to 1.7379 (at 450nm)
Refractive index : 1.3000 to 1.7100 (at 589nm)
Refractive index : 1.2912 to 1.7011 (at 680nm)
Refractive index : 1.2743 to 1.6840 (at 1100nm)
Refractive index : 1.2662 to 1.6759 (at 1550nm)

Certifications

CE Marked

Specifica

Specifica

Refractive index : ±0.0002
Refractometer, Power supply unit, Monochromatic light source device, Near infrared ray viewer, Interference filter (589nm)
100/240 V
LCD
5°C to 50°C
50/60 Hz
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali