missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angelantoni Industrie™ SterilSafe 48 Vertical Laminar Flow Biohazard Cabinet
Class II vertical laminar flow biohazard cabinet with dual motors. Angelantoni Industrie™ SterilSafe 48 Vertical Laminar Flow Biohazard Cabinet features automatic airflow adjustment.
Marca: Angelantoni Industrie™ ST-003249000
Descrizione
- Dual motors system with auto adjustment of airflow within the working space according to filter loading conditions.
- Built-in V worktop.
- Anti-blocking air return slots.
- Electrical and angled front window.
- Right side glass with three port holes to allow the insertion of facilities inside tha cabinet.
- Powerful lightning.
- Integrated UV Lamp.
- Optimal front panel angle for comfort and clear viewing.
Designed to match all GMP guidelines STERILSAFE ™ series key features includes germicidal UV lamp installed on a tilt-away holder, very bright working area, supporting structure epoxy coated, ventilation system by means of two centrifugal fans with a degree of protection IP 55, recirculation and exhaust air filtration by HEPA H14 filters according to EN1822 (with 99.995% efficiency MPPS).
Specifica
EN12469-2000, EN1822 | |
Vertical Laminar Flow Biohazard Cabinet | |
Touch Screen | |
manipulation of potentially pathogens microorganisms with low or mild risk | |
1784 mm | |
860 x 1398 x 1784 mm | |
1398 mm |
1186 mm | |
SterilSafe48 | |
HEPA | |
UV Lamp | |
1200 mm | |
860 mm |