Learn More
IKA™ HABITAT Cell, Double-walled 0.5 L Package
The HABITAT cell package for the bioreactor HABITAT research contains a 0.5 L double-walled glass vessel for cell culture applications as well as the appropriate stirring drive.
Marca:  IKA™ 0010007645
Dettagli aggiuntivi : Peso : 6.77000kg
Codice prodotto. 17705079
  • dietro preventivo / Pezzo
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
Together with the separately available control tower package (e.g. HABITAT cell) you receive all necessary components for a successful cultivation. For temperature control via the double jacket we recommend one of our our circulators such as the HRC basic or control.
Scope of delivery
  • Stand 0.5
  • HA.gv.dw.0.5 Glass vessel, double-wall
  • HA.mt.s.2 Motor, small
  • HA.hp.s.0.5 Harvest pipe, straight
  • HA.fd Feeding port (2 pcs.)
  • HA.sp.m.0.5 Micro sparger, 5 μm
  • HA.cn Condenser
  • HA.ip.pi.0.5 3-pitched blade impeller
  • HA.s.tm.1 Temperature sensor
  • HA.s.ph.0.5 pH sensor
  • HA.s.do.0.5 DO sensor
  • HA.s.fo Foam sensor
  • HA.s.lv.1 Level sensor
  • HA.cab.dw Cable and tube set
  • HA.sf.250 Sample flask (4 pcs.)
Specifica
Specifica
0.5 L
Borosilicate Glass