missing translation for 'onlineSavingsMsg'
Learn More

PSAP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10801-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897423

  • 140.00€ / 20µL
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This PSAP antibody (10801-1-AP) is expected to recognize both saposin A and B.

PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.
Specifica

Specifica

PSAP
Polyclonal
Unconjugated
PSAP
A1 activator, Cerebroside sulfate activator, Co beta glucosidase, Component C, CSAct, Dispersin, GLBA, Glucosylceramidase activator, Proactivator polypeptide, prosaposin, Protein C, PSAP, SAP 1, SAP 2 Saposin D, SAP1
Rabbit
Antigen Affinity Chromatography
RUO
19156, 5660
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P07602, Q61207
Psap
PSAP Fusion Protein Ag1194
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers