missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18423-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody should recognize saposin D.
PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.Specifica
PSAP | |
Polyclonal | |
Unconjugated | |
PSAP | |
A1 activator, Cerebroside sulfate activator, Co beta glucosidase, Component C, CSAct, Dispersin, GLBA, Glucosylceramidase activator, Proactivator polypeptide, prosaposin, Protein C, PSAP, SAP 1, SAP 2 Saposin D, SAP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19156, 25524, 5660 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07602, P10960, Q61207 | |
Psap | |
PSAP Fusion Protein Ag13266 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |