Learn More
PSMB1 Mouse, anti-Human, Unconjugated, Clone: 1C7C5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67847-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
PSMB1(Proteasome subunit beta type-1) is also named as PSC5 and belongs to the peptidase T1B family. The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The gene encodes a 241 amino acid protein with a 28 amino acid propeptide and two glycosylation sites.
Specifica
PSMB1 | |
Monoclonal | |
Unconjugated | |
150 μL | |
5689 | |
Liquid |
Western Blot, Immunohistochemistry, ELISA | |
1C7C5 | |
Mouse | |
Primary | |
Human |