Learn More
Cowie Technology™ PTFE T-Type Connector
Accepts any rigid or semi-rigid tubing. Cowie Technology™ PTFE T-Type Connector locks in place quickly with a bayonet type coupling.
Marca: Cowie Technology™ 016.1103.7.2
Dettagli aggiuntivi : Peso : 0.00900kg
Descrizione
- Bayonet fitting
- For use with rigid and semi rigid tubing such as glass, FEP or PTFE
Specifica
PTFE T-Type Connector | |
Autoclavable | |
PTFE | |
Flexible tubing in PTFE, Viton, silicone rubber or similar materials | |
1 Each | |
14 mm | |
280°C |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
6.8 mm | |
44 mm | |
-200°C | |
29 mm | |
White |