missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17551-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Differentially dephosphorylate autophosphorylated tyrosine kinases which are known to be overexpressed in tumor tissues.Specifica
PTPN18 | |
Polyclonal | |
Unconjugated | |
PTPN18 | |
BDP1, Brain derived phosphatase, PTP HSCF, PTPN18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19253, 26469, 301333 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q61152, Q99952 | |
Ptpn18 | |
PTPN18 Fusion Protein Ag11704 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |