missing translation for 'onlineSavingsMsg'
Learn More

PTPN9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 12109-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899623

  • 140.00€ / 20µL
Spedizione stimata: 20-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

MEG2 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal domain that shares a significant similarity with yeast SEC14, which is a protein that has phosphatidylinositol transfer activity and is required for protein secretion through the Golgi complex in yeast. This PTP was found to be activated by polyphosphoinositide, and is thought to be involved in signaling events regulating phagocytosis.
Specifica

Specifica

PTPN9
Polyclonal
Unconjugated
PTPN9
MEG2, PTPase MEG2, PTPMEG2, PTPN9
Rabbit
Antigen Affinity Chromatography
RUO
266611, 56294, 5780
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35239, P43378, Q641Z2
Ptpn9
PTPN9 Fusion Protein Ag2754
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali