Learn More
Imbuto filtrante Buchner in vetro Pyrex™, diametro 30 mm
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 30mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
Marca: Pyrex 3620/02
289.75 EUR valido fino al 2024-12-31
Usa il codice promo "23259" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.35000kg
Descrizione
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 30mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
- Choice of disc diameters and porosities
Specifica
35 mL | |
Pyrex Glass, Borosilicate Glass | |
For convenient filtration of solutions | |
Integrated filter plate sintered Glass disc | |
1 | |
Buchner filter funnel |
8mm | |
160 mm | |
30mm | |
100mm | |
100 to 160μm |