missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB12 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18843-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. That Rab may play a role in protein transport from recycling endosomes to lysosomes regulating, for instance, the degradation of the transferrin receptor. Involved in autophagy.Specifica
RAB12 | |
Polyclonal | |
Unconjugated | |
RAB12 | |
RAB12, Ras related protein Rab 12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
201475 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6IQ22 | |
RAB12 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |