missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB24 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11445-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
RAB24 may be involved in autophagy-related processes.Specifica
RAB24 | |
Polyclonal | |
Unconjugated | |
RAB24 | |
RAB24, Ras related protein Rab 24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19336, 53917 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35290, Q969Q5 | |
RAB24 | |
RAB24 Fusion Protein Ag1995 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |