missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11756-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Specifica
RAB2B | |
Polyclonal | |
Unconjugated | |
RAB2B | |
RAB2B, Ras related protein Rab 2B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305853, 76338, 84932 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P59279, Q8WUD1 | |
Rab2b | |
RAB2B Fusion Protein Ag2352 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |