missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABEP2 Mouse anti-Human, Rat, Clone: 1A4A4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60125-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
RABEP2, also named as RABPT5B, plays a role in membrane trafficking and in homotypic early endosome fusion. It has two isoforms with MW 59-63 kDa.Specifica
RABEP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q62835, Q9H5N1 | |
RABEP2 | |
RABEP2 Fusion Protein Ag6197 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1A4A4 | |
Unconjugated | |
RABEP2 | |
FRA, Rabaptin 5beta, RABEP2, RABPT5B | |
Mouse | |
Protein G | |
RUO | |
79874, 80754 | |
-20°C | |
Liquid |