Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Thermostatic Jacket and Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca:  Lenz Laborglasintrumente 06040279
Dettagli aggiuntivi : Peso : 0.50000kg
Codice prodotto. 10543963
  • 2427.40€ / Pezzo
Spedizione stimata: 02-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Without graduation, with laboratory flange LF, with groove
  • With two connectors (with threads GL 14 /GL 18 or ball flanges KF 15)
  • Made of DURAN™ tubing, other sizes on request
  • Delivery with 2 plastic olives GL14/18 and screw caps
Specifica
Specifica
4 L
Lab Flange 100 LF, groove and two connectors (GL 18)
1 Ea.
395 mm
236 mm