Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Thermostatic Jacket and Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 06040149
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Without graduation, with laboratory flange LF, with groove
- With two connectors (with threads GL 14 /GL 18 or ball flanges KF 15)
- Made of DURAN™ tubing, other sizes on request
- Delivery with 2 plastic olives GL14/18 and screw caps
Specifica
0.25 L | |
255 mm | |
105 mm |
Duran Glass | |
Lab Flange 60 LF, groove and two connectors (GL 14) | |
1 Ea. |