missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine IL10 / Interleukin-10 Protein
A DNA sequence encoding the Bos taurus (Bovine) IL10 / Interleukin-10, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9204-ye-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_776513.1 | |
IL10 / Interleukin-10 Protein | |
Research Use Only | |
IL10 | |
Recombinant Protein | |
Yeast | |
SRDASTLSDS SCIHLPTSLP HMLRELRAAF GKVKTFFQMK DQLHSLLLTQ SLLDDFKGYL GCQALSEMIQ FYLEEVMPQA ENHGPDIKEH VNSLGEKLKT LRLRLRRCHR FLPCENKSKA VEKVKRVFSE LQERGVYKAM SEFDIFINYI ETYMTTKMQK | |
Greater than 90% as determined by SDS-PAGE. |
281246 | |
200 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |