missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine PIGR Protein
A DNA sequence encoding the Bos taurus (Bovine) PIGR, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6494-ye-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_776568.1 | |
PIGR Protein | |
Research Use Only | |
PIGR | |
Recombinant Protein | |
Yeast | |
ESRGLIKEQY EGRLALLTEP GNGTYTVILN QLTDQDTGFY WCVTDGDTRW ISTVELKVVQ GEPSLKVPKN VTAWLGEPLK LSCHFPCKFY SFEKYWCKWS NRGCSALPTQ NDGPSQAFVS CDQNSQVVSL NLDTVTKEDE GWYWCGVKEG PRYGETAAVY VAVESRVKGS QGAKQVKAAP AGAAIQSRAG EIQNKALLDP S | |
Greater than 90% as determined by SDS-PAGE. |
281401 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |