missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Retinol-binding protein 1
A DNA sequence encoding the Bos taurus (Bovine) Retinol-binding protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9364-ye-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_776589.1 | |
Retinol-binding protein 1 | |
Research Use Only | |
RBP3 | |
Recombinant Protein | |
Yeast | |
SLGELVEGTG RLLEAHYARP EVVGQMGALL RAKLAQGAYR TAVDLESLAS QLTADLQEMS GDHRLLVFHS PGEMVAEEAP PPPPVVPSPE ELSYLIEALF KTEVLPGQLG YLRFDAMAEL ETVKAVGPQL VQLVWQKLVD TAALVVDLRY NPGSYSTAVP LLCSYFFEAE PRRHLYSVFD RATSRVTEVW TLPHVTGQRY GSHKDLYVLV SHTSGSAAEA FAHTMQDLQR ATIIGEPTAG GALSVGIYQV GSSALYASMP TQMAMSASTG EAWDLAGVEP DITVPMSVAL STARDIVTLR AKVPTVLQTA GKLVADNYAS PELGVKMAAE LSGLQSRYAR VTSEAALAEL LQADLQVLSG DPHLKTAHIP EDAKDRIPGI VPMQIPSPEV FEDLIKFSFH TNVLEGNVGY LRFDMFGDCE LLTQVSELLV EHVWKKIVHT DALIVDMRFN IGGPTSSISA LCSYFFDEGP PILLDKIYNR PNNSVSELWT LSQLEGERYG SKKSMVILTS TLTAGAAEEF TYIMKRLGRA LVIGEVTSGG CQPPQTYHVD DTDLYLTIPT ARSVGAADGS SWEGVGVVPD VAVPAEAALT RAQEMLQHT | |
Greater than 90% as determined by SDS-PAGE. |
281443 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |