missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant CMV CMV Pp28 Protein
A cDNA sequence encoding the CMV Pp28 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11463-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Pp28 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-Hcl pH 7.2, 1mM EDTA and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
CMV | |
Untagged | |
CMV Pp28 protein is >95% pure as determined by 10% PAGE (coomassie staining). |